missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Iodide Trihydrate, Extra Pure, SLR, Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Glass Jar
Specifications
Boiling Point | 1171°C |
Melting Point | 446°C |
CAS | 7790-22-9 |
Color | Beige |
Physical Form | Solid |
Quantity | 100 g |
Molecular Formula | H6ILiO3 |
MDL Number | MFCD00149770 |
Synonym | lithium iodide trihydrate, unii-ps8215ojnr, ps8215ojnr, lithium iodide trihydrate mi, lithium iodide lii , trihydrate, lithotab trihydrate iodide, i.li.3h2o, lithium iodide, reagent, lithium iodide-water 1/1/3, lithium iodide, hydrated 3h2o |
InChI Key | UMXWTWTZJKLUKQ-UHFFFAOYSA-M |
Show More |
Safety and Handling
Product Identifier
- Lithium iodide anhydrous
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'