missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium iodide, anhydrous, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Used as an electrolyte in high temperature batteries and artificial pacemakers. Phosphor , which is a complex of Iodine is used in neuron detection , in an electrolyte of the dye-sensitized solar cell. It is an electrolyte additive for lithium-sulfur batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Boiling Point | 1170°C to 1190°C |
Melting Point | 449°C |
Color | White |
Physical Form | Crystalline, Powder |
Quantity | 25 g |
Linear Formula | LiI |
Merck Index | 14,5535 |
Solubility Information | Soluble in water,ethanol and acetone. |
Formula Weight | 133.84 |
Percent Purity | 99.95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'