missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium ingot, 5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long, 99.9% (metals basis)

Brand:  Alfa Aesar™ 000658.KI

Code : NEW

Additional Details : CAS Number : 7439-93-2 Weight : 0.01000kg Transport : UN number : 1415 Chem class : 1 Pack group : I

Quantity 4 pieces

 View more versions of this product

Product Code. 11325719

  • £315.70 / Pack of 4


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Lithium ingot
5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long
UN1415
7439-93-2
MFCD00134051

Specifications

Lithium ingot
4 pieces
7439-93-2
99.9%
MFCD00134051
Odorless
Silver-white
≈116g/pc
5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long
Pieces
(metals basis)
UN1415
Vacuum can
Videos
SDS