missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium hydride, 99.4% (metals basis)

Brand:  Alfa Aesar™ 041596.22

Code : NEW

Additional Details : CAS Number : 7580-67-8 Weight : 0.10000kg Transport : UN number : 1414 Chem class : 1 Pack group : I

Quantity 100g

 View more versions of this product

Product Code. 11318728

  • £171.30 / 100g


Estimated Shipment date
from Supplier 27-04-2018
Add to basket

Specifications

Chemical Identifiers

Lithium hydride
LiH
MFCD00011074
14,5533
7580-67-8
7.95
UN1414

Specifications

Lithium hydride
Powder
(metals basis)
7.95
UN1414
0.78g/mL
Air & Moisture Sensitive
686°C
7580-67-8
99.4%
LiH
MFCD00011074
14,5533
Odorless
White to gray
100g
Videos
SDS