missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals, Puratronic™

Catalog Number p-4407566
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-24-4
FLi
25.94
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
224478
[Li+].[F-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-24-4
FLi
25.94
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
224478
[Li+].[F-]

Used in specialized UV optics. Also used as a crystal in X-ray spectrometry. Widely employed in PLED (Polymer Light Emitting Diodes) and OLED (Organic Light Emitting Diodes) as a coupling layer to enhance electron injection. Found in liquid-fluoride nuclear reactors. Used to record ionizing radiation in thermoluminescent dosimeters. Component of molten salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1680°C
Melting Point 845°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in HF. Less soluble in water. Insoluble in alcohol.
IUPAC Name lithium;fluoride
Formula Weight 25.94
Odor Odorless
Refractive Index 1.3915
Appearance White powder
Density 2.635 g/cm3 at 20°C
Chemical Name or Material Lithium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'