missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048596
Click to view available options
Quantity:
25 g
100 g
7789-24-4
FLi
25.94
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
224478
[Li+].[F-]
7789-24-4
FLi
25.94
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
224478
[Li+].[F-]

Lithium fluoride is widely used in many fields. It is used in the soldering progress for glass lining as a cosolvent. It finds application as an additive of aluminum electrolysis and rare earth electrolysis as a crystal in X-ray spectrometry. It is also used in specialized UV optics due to its large band gap and transparent to short wavelength ultraviolet radiation. It is involved to record ionizing radiation exposure from gamma rays, beta particles and neutrons in thermoluminescent dosimeters. In nuclear reactors, lithium fluoride is mixed with beryllium fluoride to form a base solvent, which is used in molten-salt reactor experiment (MSRE). Additionally, it is useful as a coupling layer to enhance electron injection in polymer light-emitting diodes (PLED) and organic light-emitting diode (OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1680°C
Melting Point 845°C
Color White
Physical Form Powder
Quantity 25 g
Linear Formula LiF
UN Number UN3288
Merck Index 14,5531
Solubility Information Soluble in water and hydrogen fluoride. Insoluble in alcohol.
IUPAC Name lithium;fluoride
Formula Weight 25.94
Percent Purity 99.98%
Odor Odorless
Refractive Index 1.3915
Density 2.635 g/mL
Assay (metals basis)
Chemical Name or Material Lithium fluoride
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'