missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 99.85% (metals basis), Thermo Scientific Chemicals

Catalog Number 11313458
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7789-24-4
FLi
25.94
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
224478
lithium;fluoride
[Li+].[F-]
This item is not returnable. View return policy
7789-24-4
FLi
25.94
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
224478
lithium;fluoride
[Li+].[F-]

Used in the preparation of hypersensitive thermoluminescent materials, dosimeters and in the room-temperature oxidation of noble metals using fluorine in anhydrous HF. It is mainly used as a component of molten salts. Lithium fluoride forms the basic constituent of the preferred fluoride salt mixture used in liquid-fluoride nuclear reactors. Lithium fluoride is widely used in PLED and OLED as a coupling layer to enhance electron injection. LiF crystals are transparent to short wavelength ultraviolet radiation, more so than any other material. LiF is therefore used in specialized UV optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1680°C
Melting Point 845°C
Color White
Physical Form Lump/Powder
Quantity 10 g
Linear Formula LiF
UN Number UN3288
Merck Index 14,5531
Solubility Information Soluble in 0.29g/100 mL (20°C) and hydrogen fluoride. Insoluble in alcohol.
Formula Weight 25.94
Percent Purity 99.85%
Odor Odorless
Refractive Index 1.3915
Density 2.635 g/mL
Assay (metals basis)
Chemical Name or Material Lithium fluoride
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'