missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecylsulfate, 20% Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
Packaging:
Nalgene bottle
Specifications
CAS | 151-21-3 |
Physical Form | Solution |
Molecular Formula | C12H25NaO4S |
Concentration | 20% Solution |
MDL Number | MFCD00036175 |
Quantity | 500 mL |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
IUPAC Name | sodium;dodecyl sulfate |
Molecular Weight (g/mol) | 288.38 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'