missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Dodecylsulfate, ≥98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Specifications
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | CH3(CH2)11OSO3Li |
UN Number | UN1325 |
IUPAC Name | lithium(1+) dodecyl sulfate |
Formula Weight | 272.33 |
Percent Purity | ≥98% |
Packaging | Plastic bottle |
Chemical Name or Material | Lithium Dodecylsulfate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'