missing translation for 'onlineSavingsMsg'
Learn More

Lithium Dodecyl Sulfate (Powder or Flakes/Molecular Biology), Fisher BioReagents

Catalog Number p-26821
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
2044-56-6
C12H25LiO4S
272.33
MFCD00007467
YFVGRULMIQXYNE-UHFFFAOYSA-M
2735071
lithium(1+) dodecyl sulfate
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
This item is not returnable. View return policy
2044-56-6
C12H25LiO4S
272.33
MFCD00007467
YFVGRULMIQXYNE-UHFFFAOYSA-M
2735071
lithium(1+) dodecyl sulfate
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
Lithium Dodecyl Sulfate (LDS) is a detergent used for dissociating proteins.
TRUSTED_SUSTAINABILITY
Absorbance 0.01 max. (3% solution) at 405nm, 0.05 max. (3% solution) at 280nm, 0.100 AU max. at 260nm, 0.3 max. (3% solution) at 230nm
Color White
Physical Form Solid
Assay Percent Range ≥99 %
Linear Formula CH3(CH2)11OSO3Li
Quantity 25 g
Identification Pass Test
Formula Weight 273.18
Percent Purity ≥99% (≥98% C12-OH)
Grade Molecular Biology
Packaging Glass Bottle
Chemical Name or Material Lithium Dodecyl Sulfate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium dodecyl sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'