missing translation for 'onlineSavingsMsg'
Learn More

Lithium DL-2-aminoadipate, 97+%, Thermo Scientific™

Catalog Number p-7059530
Click to view available options
Quantity:
0.1 g
0.5 g
2 g
This item is not returnable. View return policy
C6H9Li2NO4
MFCD06657665
DL-2-Aminoadipic acid lithium salt
This item is not returnable. View return policy
C6H9Li2NO4
MFCD06657665
DL-2-Aminoadipic acid lithium salt

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Linear Formula [O2C(CH2)3CH(NH2)CO2]Li2
Quantity 0.1 g
Formula Weight 173.02
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Lithium DL-2-aminoadipate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'