missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium dimethylamide, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is used in the synthesis of some d0 organoimido complexes of the early transition metals and in meta rearrangement in the reaction of triphenyl-(p-bromophenyl)-silane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | White |
Odor | Amine-like |
Linear Formula | LiN(CH3)2 |
Quantity | 5 g |
UN Number | UN3393 |
Beilstein | 3618233 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts violently with water. |
Formula Weight | 51.02 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'