missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium dimethylamide, 10% w/v in hexane

Brand:  Alfa Aesar™ 045491.AK

Code : NEW

Additional Details : CAS Number : 3585-33-9 Weight : 0.00025kg Transport : UN number : 3394 Chem class : 1 Pack group : I

Quantity 250mL

 View more versions of this product

Product Code. 15442307

  • £153.49 / 250mL


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Chemical Identifiers

Lithium dimethylamide
C2H6LiN
51.02
UN3394
3585-33-9
LiN(CH3)2
MFCD00008295

Specifications

Lithium dimethylamide
Slurry
LiN(CH3)2
MFCD00008295
10% w/v in hexane
250mL
Air & Moisture Sensitive
3585-33-9
C2H6LiN
51.02
UN3394
White
Petrol-like
Videos
SDS