missing translation for 'onlineSavingsMsg'
Learn More

Lithium deuteride, 98%(Isotopic), Alfa Aesar™

Used in thermonuclear Fusion

Brand:  Alfa Aesar 044100.SS

Code : NEW

Additional Details : CAS Number : 13587-16-1 Weight : 0.00020kg Transport : UN number : 1414 Chem class : 1 Pack group : I

Quantity 0.2g

 View more versions of this product

Product Code. 11329568

  • £19.82 / 0.20g


Estimated Shipment date
from Supplier 21-05-2018
Add to basket

Specifications

Chemical Identifiers

Lithium deuteride
Powder
DLi
MFCD00011091
13587-16-1
98% (Isotopic)
8.95

Specifications

Lithium deuteride
Powder
DLi
8.95
200°C (392°F)
Gray
13587-16-1
98% (Isotopic)
Gray powder
MFCD00011091
Odorless
0.2g
Videos
SDS