missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium cobalt(III) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Lithium cobalt(III) oxide is used in the positive electrodes of lithium-ion batteries and advanced batteries. It acts as a starting material for secondary battery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Blue |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.5% (Metal basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 97.88 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | Dark blue powder |
Chemical Name or Material | Lithium cobalt(III) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'