missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium chloride hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals
83.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Lithium chloride is widely employed in lithium metal production. It is used as a brazing flux for aluminum in automobile parts. Its use can also be seen as a desiccant for drying air streams. In biochemical applications, lithium chloride is used to precipitate RNA from cellular extracts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >98°C-H2O |
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 25 g |
Assay Percent Range | 99.996% (Metals basis) |
Solubility Information | Soluble in water. |
IUPAC Name | lithium;chloride;hydrate |
Formula Weight | 42.39 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'