missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium chloride, 200mM aq. soln., Thermo Scientific™
Click to view available options
Quantity:
100 mL
250 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Used in formation of an active Mn(0) species useful for radical cyclization reactions. It is utilized in a variety of assays to study cell-fate and neurobiology. In developing Xenopus embryos, lithium chloride is observed to exert inhibition of GSK-3β (glycogen synthase kinase-3β). Additionally, in transfected Chinese Hamster Ovary cells (CHO) stimulated with angiotension II (Ang II), it was noted that lithium chloride enhanced the production of inositol triphosphate. The antiviral properties of lithium chloride were noted in a study which showed that the compound inhibited pseudorabis virus infection in vitro. It is primarily used in the synthesis of lithium metal by electrolysis. Also acts as a prescribed drug for bipolar mood disorder, it may also exhibit anti-inflammatory properties.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, ether, pyridine, nitrobenzene, acetone, and amyl alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Used in formation of an active Mn(0) species useful for radical cyclization reactions. It is utilized in a variety of assays to study cell-fate and neurobiology. In developing Xenopus embryos, lithium chloride is observed to exert inhibition of GSK-3β (glycogen synthase kinase-3β). Additionally, in transfected Chinese Hamster Ovary cells (CHO) stimulated with angiotension II (Ang II), it was noted that lithium chloride enhanced the production of inositol triphosphate. The antiviral properties of lithium chloride were noted in a study which showed that the compound inhibited pseudorabis virus infection in vitro. It is primarily used in the synthesis of lithium metal by electrolysis. Also acts as a prescribed drug for bipolar mood disorder, it may also exhibit anti-inflammatory properties.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, ether, pyridine, nitrobenzene, acetone, and amyl alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | LiCl |
Solubility Information | Not miscible or difficult to mix in water. Soluble in alcohol,ether,pyridine,nitrobenzene,acetone,and amyl alcohol. |
IUPAC Name | lithium(1+) chloride |
Formula Weight | 42.39 |
Concentration or Composition (by Analyte or Components) | 200mM aq. soln. |
Odor | Odorless |
Chemical Name or Material | Lithium chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'