missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Carbonate, purum p.a, ≥98.0% (T), Honeywell™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Boiling Point | 1300°C |
Melting Point | 720°C |
Color | White |
pH | 9 to 11 (at 1g/L, 20°C) |
Physical Form | Crystalline |
Quantity | 1 kg |
Linear Formula | Li2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 3999191 |
Formula Weight | 73.89g/mol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'