missing translation for 'onlineSavingsMsg'
Learn More

Lithium carbonate, 99%, Thermo Scientific Chemicals

Catalog Number p-4407559
92.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
500 g
2 kg
10 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
554-13-2
CLi2O3
73.89
MFCD00011084
XGZVUEUWXADBQD-UHFFFAOYSA-L
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
11125
CHEBI:6504
[Li+].[Li+].[O-]C([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
554-13-2
CLi2O3
73.89
MFCD00011084
XGZVUEUWXADBQD-UHFFFAOYSA-L
lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
11125
CHEBI:6504
[Li+].[Li+].[O-]C([O-])=O

Lithium carbonate-based glasses are used in overware. Used in the manufacture of lithium-ion battery cathodes. Pharmaceutical composition of lithium carbonate has been shown to exhibit antimanic effects specifically to treat mania associated with bipolar disorder. Used in both low-fire and high-fire ceramic glaze, tile adhesives, and in the processing of metal oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1310°C(decomposition)
Melting Point 723°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99%
Solubility Information Soluble in water and dilute acids. Insoluble in alcohol.
IUPAC Name dilithium(1+) carbonate
Formula Weight 73.9
Odor Odorless
Appearance White powder
Density 2.11 g/cm3 at 20°C
Chemical Name or Material Lithium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'