missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Employed as adsorbents, functional fluids (closed systems) and intermediates. Used as an operating medium for air-conditioning and industrial drying system. It is also used in manufacturing pharmaceuticals and alkylation process. It is used as brazing and welding fluxes. Used for reconstitution of brines, as swelling agent for proteins, as electrolyte component in lithium batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1265°C |
Melting Point | 550°C |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.998% (Metals basis) |
Solubility Information | Soluble in water (at 20°C 1490g/L),alcohol and glycol. |
Formula Weight | 86.85 |
Sensitivity | Hygroscopic |
Density | 3.464 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'