missing translation for 'onlineSavingsMsg'
Learn More

Lithium bromide, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11320368
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7550-35-8
BrLi
86.844
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]
This item is not returnable. View return policy
7550-35-8
BrLi
86.844
MFCD00011077
AMXOYNBUYSYVKV-UHFFFAOYSA-M
lithium bromide, lithium monobromide, lithium bromide libr, lithiumbromide, libr, lithium 1+ ion bromide, lithium bromide, anhydrous, lithium bromide, ultra dry, bromolithium, lithium-bromide
82050
CHEBI:63042
lithium;bromide
[Li+].[Br-]

Employed as adsorbents, functional fluids (closed systems) and intermediates. Used as an operating medium for air-conditioning and industrial drying system. It is also used in manufacturing pharmaceuticals and alkylation process. It is used as brazing and welding fluxes. Used for reconstitution of brines, as swelling agent for proteins, as electrolyte component in lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1265°C
Melting Point 550°C
Color White
Physical Form Powder or Granules
Quantity 25 g
Linear Formula LiBr
Merck Index 14,5526
Solubility Information Soluble in water (at 20°C 1490g/L),alcohol and glycol.
Formula Weight 86.85
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.464 g/mL
Assay (metals basis)
Chemical Name or Material Lithium bromide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'