Learn More
Lithium borohydride, 95%, Thermo Scientific Chemicals
Descripción
Strong reducing agent. Used to reduce compounds containing ketones, aldehydes, ester carbonyls, acid chlorides, lactones, and epoxidesLithium borohydride is a versatile reducing agent for aldehydes, ketones, nitriles, primary amides, acid chlorides, lactones, epoxides, and esters. It is involved in the preparation of intercalation compounds and nanocomposites. It is used as a precursor for other borohydrides and acts as a catalyst in hydroboration reactions. It is known to be good reversible storage compound for anhydrous ammonia and hydrogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Strong reducing agent
- Used to reduce compounds containing ketones, aldehydes, ester carbonyls, acid chlorides, lactones, and epoxides
Especificaciones
Melting Point | ∼280°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Linear Formula | LiBH4 |
UN Number | UN1413 |
Merck Index | 14,5525 |
Solubility Information | Soluble in ether,THF,and aliphatic aminesSoluble in ether,tetrahydrofuran,aliphatic amines and ethanol. |
Formula Weight | 21.78 |
Percent Purity | 95% |
Mostrar más |
Seguridad y manipulación
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH014-Reacts violently with water.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only