missing translation for 'onlineSavingsMsg'
Learn More

Lithium bis(trimethylsilyl)amide, 15% in 2-MeTHF/Ethylbenzene, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 446111000

Code : 29

Additional Details : CAS Number : 4039-32-1 Weight : 0.13560kg Transport : UN number : 2924 Chem class : 1 Pack group : II

Quantity 100mL
Packaging AcroSeal™ Glass bottle

 View more versions of this product

Product Code. 12317173

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Lithium bis(trimethylsilyl)amide
15%
[(CH3)3Si]2NLi
100-41-4
C6H18LiNSi2
167.33

Specifications

Lithium bis(trimethylsilyl)amide
15%
[(CH3)3Si]2NLi
0.8560g/mL
Solubility in water: decomposes.
100mL
100-41-4
C6H18LiNSi2
167.33
AcroSeal™ Glass bottle
0.856
−11°C
Videos
SDS