missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum hydride, powder, 97%, Thermo Scientific™

Catalog Number p-7023999
Click to view available options
Quantity:
10 g
25 g
100 g
1000 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
16853-85-3
AlH4Li
37.95
MFCD00011075
OCZDCIYGECBNKL-UHFFFAOYSA-N
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
21226445
[Li+].[AlH4-]
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
16853-85-3
AlH4Li
37.95
MFCD00011075
OCZDCIYGECBNKL-UHFFFAOYSA-N
lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
21226445
[Li+].[AlH4-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Lithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium, which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.

Solubility
Soluble in water, diethyl ether and terahydrofuran.

Notes
Air and moisture sensitive. Incompatible with water, strong oxidizing agents, alcohols, carboxylic acid, peroxides, chlorinated solvents, halogens and oxygen. It reacts violently with water and release hydrogen gas.
TRUSTED_SUSTAINABILITY
Melting Point >125°C (decomposition)
Physical Form Powder
Quantity 10 g
UN Number UN1410
Merck Index 14,349
Solubility Information Soluble in water,diethyl ether and terahydrofuran.
IUPAC Name aluminum;lithium;hydride
Formula Weight 37.95
Percent Purity 97%
Odor Odorless
Sensitivity Moisture sensitive
Density 0.917
Chemical Name or Material Lithium aluminum hydride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'