missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium aluminum hydride, 10% in THF

Reducing agent for pharmaceuticals and organic chemicals

Brand:  Alfa Aesar™ 045502.AP

Code : NEW

Additional Details : CAS Number : 16853-85-3 Weight : 0.00050kg Transport : UN number : 3399 Chem class : 4.3 Pack group : I

Quantity 500mL

Product Code. 15412327

  • £249.10 / 500mL


EU Stock 1
Add to basket

Specifications

Specifications

Lithium aluminum hydride
Liquid
LiAlH4
MFCD00011075
14,349
0.90g/mL
Air & Moisture Sensitive
500mL
16853-85-3
H4AlLi
37.95
UN3399
10% in THF
Characteristic
Pale gray
Videos
SDS