missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminium hydride, 95%, in resealable tin cans, ACROS Organics™

Brand:  Acros Organics 332100100

Code : 29

Additional Details : CAS Number : 16853-85-3 Weight : 0.06000kg Transport : UN number : 1410 Chem class : 4.3 Pack group : I

Quantity 10g
Packaging Plastic bag in resealable tin can

 View more versions of this product

Product Code. 10499502

  • £21.30 / 10g


EU Stock 181
Add to basket

Description

Description

Reduction reagent
Specifications

Chemical Identifiers

Lithium aluminium hydride
16853-85-3
95%
LiAlH4
MFCD00011075
In Resealable Tin Cans
95%
AlH4Li
37.95
Lithium tetrahydridoaluminate, LAH

Specifications

Lithium aluminium hydride
16853-85-3
95%
AlH4Li
37.95
01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Lithium tetrahydridoaluminate, LAH
Plastic bag in resealable tin can
White to light gray
10g
In Resealable Tin Cans
Crystalline Powder
95%
LiAlH4
MFCD00011075
15, 344
0.9
Solubility in water: reacts. Other solubilities: soluble in diethyl ether (39g/100g at 25 °C), thf/, toluene (70:30) (17g/100g at 25 °C), in thf/mtbe (, 25:75) (12g/100g at 25 °C), in thf/toluene (15:85), (5g/100g at 25 °C);, practically insoluble in hyd
125.0°C
Videos
SDS