missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049128
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
546-89-4
C2H3LiO2
65.98
MFCD00013057
XIXADJRWDQXREU-UHFFFAOYSA-M
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
3474584
CHEBI:63045
[Li+].CC([O-])=O
This item is not returnable. View return policy
546-89-4
C2H3LiO2
65.98
MFCD00013057
XIXADJRWDQXREU-UHFFFAOYSA-M
lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
3474584
CHEBI:63045
[Li+].CC([O-])=O

Lithium acetate hydrate is used in the laboratory as buffer for gel electrophoresis of DNA and RNA. It is also used to permeabilize the cell wall of yeast for use in DNA transformation. It is a metal acetate for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 57.8°C
Physical Form Crystalline
Quantity 5 g
Merck Index 14,5521
Solubility Information Soluble in water.
IUPAC Name lithium(1+) acetate
Formula Weight 65.98 (Anhydrous)
Percent Purity 99.998%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Lithium acetate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'