missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate dihydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog Number p-44212
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
6108-17-4
C2H3LiO2·2H2O
102.02
MFCD00066949
IAQLJCYTGRMXMA-UHFFFAOYSA-M
lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
23666338
lithium;acetate;dihydrate
[Li+].CC(=O)[O-].O.O
6108-17-4
C2H3LiO2·2H2O
102.02
MFCD00066949
IAQLJCYTGRMXMA-UHFFFAOYSA-M
lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
23666338
lithium;acetate;dihydrate
[Li+].CC(=O)[O-].O.O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 56°C to 58°C
Color White
pH 7.5 to 9.5 (5% soln.)
Physical Form Adhering Crystalline Powder and Chunks
Quantity 250 g
Assay Percent Range 98%
Linear Formula CH3CO2Li·2H2O
Beilstein 02, 107
Merck Index 15, 5577
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: very soluble in hot water,215g/L ethanol
Formula Weight 102.02
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Lithium acetate dihydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Lithium acetate dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Acetate, lithium, dihydrate, Acetic acid, lithium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'