missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium 2-[(4-methylpiperazin-1-yl)methyl]benzoate, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 915707-44-7 |
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Molecular Formula | C13H17LiN2O2 |
MDL Number | MFCD09064954 |
Quantity | 1 g |
Synonym | lithium 2-4-methylpiperazin-1-yl methyl benzoate, lithium 1+ ion 2-4-methylpiperazin-1-yl methyl benzoate, lithotab 2-4-methylpiperazin-1-yl methyl benzoate, 4-methylpiperazin-1-yl methyl benzoic acid, lithium salt, 2-4-methylpiperazinyl methyl benzoic acid, lithium salt, benzoicacid,2-4-methyl-1-piperazinyl methyl-,lithium salt 1:1, lithium 1+ 2-4-methylpiperazin-1-yl methyl benzoate |
InChI Key | BHMRBFXQIGCNNV-UHFFFAOYSA-M |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'