missing translation for 'onlineSavingsMsg'
Learn More

Statebourne Cryogenics™ Liquid Dispenser Devices

Brand:  Statebourne Cryogenics™ 9701157

Code : 04

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 11417376

  • £1184.63 / Each


Estimated Shipment
16-07-2018
Add to basket

Description

Description

  • Pressure dispenser
  • Durable and lightweight
Specifications

Specifications

Liquid Withdrawal Device
51mm
5 Years
2 to 50L Models.
Videos