missing translation for 'onlineSavingsMsg'
Learn More

Statebourne Cryogenics™ Liquid Dispenser Devices

Brand:  Statebourne Cryogenics™ 9701056

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11939384

  • £1116.21 / Each


Estimated Shipment
21-06-2018
Add to basket

Description

Description

Comprehensive range of accessories available Series of liquid nitrogen dewars providing a range of capacities from 2L to 50L to suit individual laboratory storage and liquid handling requirements.
  • All aluminium construction
  • All units guaranteed for three years
  • Low pressure dispenser on models CL20 to 50
  • ISO 9001 designed and manufactured
  • Supplied with neckplugs and dipstick
Specifications

Specifications

Liquid Withdrawal Device
51mm
5 Years
5, 10, 20 and 25L models.
Videos
Documentation