missing translation for 'onlineSavingsMsg'
Learn More

Statebourne Cryogenics™ Liquid Dispenser Devices

Brand:  Statebourne Cryogenics™ 9701152

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11407376

  • £1171.55 / Each


Estimated Shipment
21-05-2018
Add to basket

Specifications

Specifications

Liquid Withdrawal Device
63mm
5 Years
2 to 50L Models.
Videos
Documentation