missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lipocalin-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Lipocalin-1 |
---|---|
Clone | 223908 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Lipocalin-1 Monoclonal specifically detects Lipocalin-1 in Human samples. It is validated for Western Blot.Specifications
Lipocalin-1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P31025 | |
3933 | |
Mouse myeloma cell line NS0-derived recombinant human Lipocalin-1 His19-Asp176 Accession # P31025 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Lipocalin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Lipocalin-2. |
223908 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LCN1, lipocalin 1 (tear prealbumin), lipocalin 1-like 2, lipocalin-1, Lipocalin1, MGC71975, PMFA, Tear lipocalin, Tear prealbumin, Tlc, TPVon Ebner gland protein, VEG protein, VEGP, VEGPlipocalin 1 (protein migrating faster than albumin, tear prealbumin) | |
LCN1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only