missing translation for 'onlineSavingsMsg'
Learn More

Linomide, Tocris Bioscience™

Immunomodulator with antiangiogenic properties

Brand:  Tocris (Bio-Techne) 146150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13469543

  • £496.86 / 50mg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunomodulator with stimulatory and antitumor properties. Inhibits in vivo apoptosis of T cells and inhibits angiogenesis in rats in vivo.
Specifications

Chemical Identifiers

84088-42-6
308.337
linomide, roquinimex, roquinimexum latin, roquinimex usan:inn, ccris 7673, fcf 89, fcf-89, n-phenyl-n-methyl-1,2-dihydro-4-hydroxy-1-methyl-2-oxo-quinoline-3-carboxamide, 1,2-dihydro-4-hydroxy-n,1-dimethyl-2-oxo-3-quinolinecarboxanilide, 4-hydroxy-n,1-dimethyl-2-oxo-n-phenyl-1,2-dihydroquinoline-3-carboxamide
4-hydroxy-N,1-dimethyl-2-oxo-N-phenylquinoline-3-carboxamide
C18H16N2O3
SGOOQMRIPALTEL-UHFFFAOYSA-N
54676478
CN1C2=CC=CC=C2C(=C(C1=O)C(=O)N(C)C3=CC=CC=C3)O

Specifications

50mg
84088-42-6
linomide, roquinimex, roquinimexum latin, roquinimex usan:inn, ccris 7673, fcf 89, fcf-89, n-phenyl-n-methyl-1,2-dihydro-4-hydroxy-1-methyl-2-oxo-quinoline-3-carboxamide, 1,2-dihydro-4-hydroxy-n,1-dimethyl-2-oxo-3-quinolinecarboxanilide, 4-hydroxy-n,1-dimethyl-2-oxo-n-phenyl-1,2-dihydroquinoline-3-carboxamide
CN1C2=CC=CC=C2C(=C(C1=O)C(=O)N(C)C3=CC=CC=C3)O
308.337
308.33
Linomide
C18H16N2O3
SGOOQMRIPALTEL-UHFFFAOYSA-N
4-hydroxy-N,1-dimethyl-2-oxo-N-phenylquinoline-3-carboxamide
54676478
>99%
Videos
Documents