missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LINGO-2 Mouse anti-Human, Clone: 382008, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | LINGO-2 |
---|---|
Clone | 382008 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human LINGO-2 Cys28-Leu542 Accession # Q7L985 |
Gene ID (Entrez) | 158038 |
Gene Accession No. | Q7L985 |
Description
LINGO-2 Monoclonal antibody MAB3679SP Specifically detects LINGO-2 in Human samples. It is validated for Western BlottingSpecifications
LINGO-2 | |
Mouse myeloma cell line NS0-derived recombinant human LINGO-2 Cys28-Leu542 Accession # Q7L985 | |
Q7L985 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
382008 | |
158038 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human LINGO-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LINGO-1 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only