Learn More
Laboratoires Anios™ Wipes LINGET’ANIOS
Wipes LINGET’ANIOS are impregnated with a cleaning and disinfecting solution, for the treatment non-immersible and non-invasive medical devices between two patients. Pleasantly scented.
Brand: Laboratoires Anios 299421
Code : NEW
Additional Details : Weight : 3.78000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
CLEANING AND DISINFECTING IMPREGNATION SOLUTION
The impregnation solution of LINGET’ANIOS is :
- Limpid colorless
- Density at +20°C : ≈ 0.934
- pH : ≈ 6
Specifications
Linget’Anios Wipers | |
Standard EN 1275 Candida albicans | |
6.5L x 10 cmW | |
1-ply |
Safety and Handling
- LINGET'ANIOS
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403-Store in a well-ventilated place.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ETHYL ALCOHOL, PROPAN-2-OL, D-GLUCONIC ACID, COMPOUND WITHN, N''-BIS(4-CHLOROPHENYL)-3, 12- DIIMIN