missing translation for 'onlineSavingsMsg'
Learn More

Linalool, 97%, Thermo Scientific Chemicals

Catalog Number 10464975
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
78-70-6
C10H18O
154.25
MFCD00008906
CDOSHBSSFJOMGT-UHFFFAOYSA-N
linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
6549
CHEBI:17580
3,7-dimethylocta-1,6-dien-3-ol
CC(=CCCC(C)(C=C)O)C
78-70-6
C10H18O
154.25
MFCD00008906
CDOSHBSSFJOMGT-UHFFFAOYSA-N
linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
6549
CHEBI:17580
3,7-dimethylocta-1,6-dien-3-ol
CC(=CCCC(C)(C=C)O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8610g/mL
Boiling Point 199.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
Refractive Index 1.4600 to 1.4640
Quantity 2.5 L
Beilstein 01, 462
Merck Index 15, 5550
Specific Gravity 0.861
Specific Rotation -2° to +2° (20°C, 589nm) (neat)
Solubility Information Solubility in water: 1.45g/L (25°C). Other solubilities: miscible with alcohol and ether
Viscosity 4.4 mPa.s (25°C)
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Linalool, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'