missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563557
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KRLKKLSELT SRKAQPKATD LNSA Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.
LIMS2 Polyclonal antibody specifically detects LIMS2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
LIMS2 | |
Polyclonal | |
Unconjugated | |
Lims2 | |
ILK-binding protein; LGMD2W; LIM and senescent cell antigen like domains 2; LIM and senescent cell antigen-like domains 2; LIM and senescent cell antigen-like-containing domain protein 2; LIM zinc finger domain containing 2; LIM-like protein 2; Lims2; LIM-type zinc finger domains 2; Particularly interesting new Cys-His protein 2; Pinch2; PINCH-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55679 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z4I7 | |
Lims2 | |
Recombinant protein corresponding to Human LIMS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only