missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM1 Mouse anti-Human, Clone: 320416, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | LIM1 |
---|---|
Clone | 320416 |
Immunogen | E. coli-derived recombinant human LIM1 Leu265-Trp406 Accession # P48742 |
Gene ID (Entrez) | 3975 |
Gene Accession No. | P48742 |
Description
LIM1 Monoclonal specifically detects LIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LIM1 | |
E. coli-derived recombinant human LIM1 Leu265-Trp406 Accession # P48742 | |
P48742 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
320416 | |
3975 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human LIM1 in direct ELISAs and Western blots. | |
Unconjugated | |
Western Blot, Immunohistochemistry | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only