missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIGHT/TNFSF14 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | LIGHT/TNFSF14 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse LIGHT (R&D Systems, Catalog # 1794-LT) Asp72-Val239 Accession # Q9QYH9 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
LIGHT/TNFSF14 Polyclonal specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Western Blot.Specifications
LIGHT/TNFSF14 | |
Goat | |
Polyclonal | |
IgG | |
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot | |
RUO |
Mouse myeloma cell line NS0-derived recombinant mouse LIGHT (R&D Systems, Catalog # 1794-LT) Asp72-Val239 Accession # Q9QYH9 | |
Primary | |
Mouse | |
Unconjugated | |
8740 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only