missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel HPLC column 250mm x 4mm I.D. analytical LiChrospher 60 RP Select B particle size 5µm

Brand:  Macherey Nagel 728038.40

HPLC column 250mm x 4mm I.D. analytical LiChrospher 60 RP Select B particle size 5µm

Code : 16

Additional Details : Weight : 0.01000kg

Product Code. 10439171

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.