missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel HPLC column 250mm x 3mm I.D. analytical LiChrospher 60 RP Select B particle size 5µm

Brand:  Macherey Nagel 728038.30

HPLC column 250mm x 3mm I.D. analytical LiChrospher 60 RP Select B particle size 5µm

Code : 16

Additional Details : Weight : 0.01000kg

Product Code. 10286611

  • £655.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket