missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel HPLC column 125mm x 3mm I.D. analytical LiChrospher 100 RP 8 ec particle size 5µm

Brand:  Macherey Nagel 728028.30

HPLC column 125mm x 3mm I.D. analytical LiChrospher 100 RP 8 ec particle size 5µm

Code : 16

Additional Details : Weight : 0.01000kg

Product Code. 10650732

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.