missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ LiChrospher™ 100 RP 8 CC Guard Columns

Brand:  Macherey-Nagel™ 728051.30

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12726561

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

For HPLC
Octyl
Not Encapped
5μm
Acetonitrile/Water
HPLC
3mm
LiChrospher® RP 8
12.5%
L7
Acetonitrile/Water
100Å
8mm
LiChrospher® RP 8
CC Guard Column
Guard Column
Videos