missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ LiChrospher™ 100 RP 8 Analytical HPLC Column

Brand:  Macherey-Nagel™ 728026.20

Code : 16

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10114901

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

12.5%
Octyl
No
Acetonitrile/water
5μm
100Å
Acetonitrile/water
L7
LiChrospher® RP 8
Analytical
2mm
250mm
LiChrospher® RP 8
LiChrospher® RP 8
1Pack
Analytical HPLC column
HPLC
Videos