missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel HPLC column 125mm x 3mm I.D. analytical LiChrospher 100 RP 8 particle size 5µm

Brand:  Macherey Nagel 728025.30

HPLC column 125mm x 3mm I.D. analytical LiChrospher 100 RP 8 particle size 5µm

Code : 16

Additional Details : Weight : 0.01000kg

Product Code. 10766332

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.