missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel HPLC guard column 2mm I.D. analytical LiChrospher 100 RP 18 ec particle size 5µm

Brand:  Macherey Nagel 728054.30

HPLC guard column 2mm I.D. analytical LiChrospher 100 RP 18 ec particle size 5µm

Code : 16

Additional Details : Weight : 0.50000kg

Product Code. 12782093

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.