missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ LiChrospher™ 100 RP 18 ec CC Guard Columns

Brand:  Macherey-Nagel™ 728054.40

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12792093

  • £168.00 / Pack of 3


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

For HPLC
Octadecyl
Yes
5μm
Acetonitrile/Water
HPLC
4mm
LiChrospher® RP 18 ec
21%
L1
Acetonitrile/Water
100Å
8mm
LiChrospher® RP 18 ec
CC Guard Column
Guard Column
Videos