missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel™ LiChrospher™ 100 RP 18 Analytical HPLC Column

Brand:  Macherey Nagel™ 728032.40

Code : 16

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10679191

  • £446.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

21%
Octadecyl
No
Acetonitrile/water
5μm
100Å
Acetonitrile/water
L1
LiChrospher® RP 18
Analytical
4mm
250mm
LiChrospher® RP 18
LiChrospher® RP 18
1Pack
Analytical HPLC column
HPLC
Alternative Products