Learn More
Tocris Bioscience™ Licarbazepine
Active metabolite of oxcarbazepine (Cat. No. 3864)
Brand:  Tocris Bioscience™ 386510
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443724
  • £124.20 / 10mg
Estimated Shipment
28-06-2019
Description
Description
Active metabolite of oxcarbazepine (Cat. No. 3864). Produces dose-dependent inhibition of glutamatergic excitatory postsynaptic potentials (EPSPs). Displays anticonvulsant activity; exhibits minor potentiation of GABAA receptor currents.
Specifications
Specifications
Licarbazepine
>99
10mg
Licarbazepine
254.28